7-异丙氧基-3-苯基-4H-1-苯并吡喃-4-酮 ≥98.0% (by GC)
供应商: TCI
Synonyms:
(依普黄酮)
I0669-25G
I0669-5G
TCIAI0669-25GEA
0
CNY
TCIAI0669-25G
TCIAI0669-5G
7-异丙氧基-3-苯基-4H-1-苯并吡喃-4-酮 ≥98.0% (by GC)
7-异丙氧基-3-苯基-4H-1-苯并吡喃-4-酮
分子式:
C₁₈H₁₆O₃ |
MDL编号:
MFCD00221719 CAS号: 35212-22-7 Merck Index: 14,05074 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 115.0 to 119.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...