Ethyl Indoline-2-carboxylate ≥95.0% (by GC)

供应商: TCI

Synonyms: Ethyl indoline-2-carboxylate, Indoline-2-carboxylic Acid Ethyl Ester

I0595-25G I0595-5G
TCIAI0595-25GEA 0 CNY
TCIAI0595-25G TCIAI0595-5G
Ethyl Indoline-2-carboxylate ≥95.0% (by GC)
Ethyl Indoline-2-carboxylate
分子式: C₁₁H₁₃NO₂
分子量: 191.23 g/mol
存储条件: Refrigerator
MDL编号: MFCD06797114
CAS号: 50501-07-0

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 53.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR