顺式-3-碘丙烯酸乙酯 ≥98.0% (by GC) stabilised

供应商: TCI
Warning

Synonyms: (Z)-Ethyl 3-iodoacrylate, cis-3-Iodoacrylic Acid Ethyl Ester

Stabilization: Stabilised with copper chip

I0502-25G I0502-5G
TCIAI0502-25GEA 0 CNY
TCIAI0502-25G TCIAI0502-5G
顺式-3-碘丙烯酸乙酯 ≥98.0% (by GC) stabilised
顺式-3-碘丙烯酸乙酯
分子式: C₅H₇IO₂
分子量: 226.01 g/mol
闪点: 99°C
存储条件: Refrigerator
MDL编号: MFCD00209584
CAS号: 31930-36-6

现在订购

产品技术规格

Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR