4-Penten-1-yl isothiocyanate ≥95.0% (by GC)

供应商: TCI
Danger

Synonyms: Isothiocyanic Acid 4-Penten-1-yl Ester

I0444-1G I0444-5G
TCIAI0444-1GEA 0 CNY
TCIAI0444-1G TCIAI0444-5G
4-Penten-1-yl isothiocyanate ≥95.0% (by GC)
4-Penten-1-yl isothiocyanate
分子式: C₆H₉NS
分子量: 127.21 g/mol
沸点: 194°C
MDL编号: MFCD00191527
CAS号: 18060-79-2
UN号: 1992
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e42ec11b1950c509fb3c7ca2acdd059e5b19fa55&vid=e42ec11b1950c509fb3c7ca2acdd059e5b19fa55&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR