间苯二甲酸二苯酯 ≥99.0% (by GC)

供应商: TCI
Warning

Synonyms: Isophthalic Acid Diphenyl Ester

I0416-10G I0416-25G
TCIAI0416-10GEA 0 CNY
TCIAI0416-10G TCIAI0416-25G
间苯二甲酸二苯酯 ≥99.0% (by GC)
间苯二甲酸二苯酯
分子式: C₂₀H₁₄O₄
分子量: 318.33 g/mol
MDL编号: MFCD00046051
CAS号: 744-45-6
EINECS: 212-014-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 136.0 to 139.0 deg-C
Solubility in Acetone almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR