Sodium-1-propanesulphonate ≥98.0% (by titrimetric analysis) for ion pair chromatography

供应商: TCI

Synonyms: Propanesulphonic acid sodium salt, 1-Propanesulphonic acid Sodium Salt, Propanesulphonic acid Sodium Salt, IPC-ALKS-3

I0341-25G I0341-5G
TCIAI0341-25GEA 0 CNY
TCIAI0341-25G TCIAI0341-5G
Sodium-1-propanesulphonate ≥98.0% (by titrimetric analysis) for ion pair chromatography
Sodium-1-propanesulphonate
分子式: C₃H₈O₃S
分子量: 146.14 g/mol
MDL编号: MFCD00062546
CAS号: 14533-63-2
EINECS: 238-564-7

现在订购

产品技术规格

Appearance White powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Absorbance (210nm) max. 1.000
Absorbance (230nm) max. 0.300
Absorbance (254nm) max. 0.130


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR