Diethyl isobutylmalonate ≥98.0% (通过GC)

供应商: TCI

I0251-25ML I0251-500ML
TCIAI0251-25MLEA 0 CNY
TCIAI0251-25ML TCIAI0251-500ML
Diethyl isobutylmalonate ≥98.0% (通过GC)
Diethyl isobutylmalonate
分子式: C₁₁H₂₀O₄
沸点: 225°C
MDL编号: MFCD00026869
CAS号: 10203-58-4
EINECS: 233-504-6

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR