Isopropyl phosphate (mono and di ester mixture)
供应商: TCI
|
Danger
|
Synonyms:
Phosphoric Acid Isopropyl Ester
I0247-25ML
I0247-500ML
TCIAI0247-25MLEA
0
CNY
TCIAI0247-25ML
TCIAI0247-500ML
Isopropyl phosphate (mono and di ester mixture)
Isopropyl phosphate (mono and di ester mixture)
分子式:
C₆H₁₅O₄P 分子量: 182.16 g/mol |
MDL编号:
MFCD00135965 UN号: 1793 ADR: 8 III |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Assay of mono ester | 25.0 to 45.0 % |
Assay of Diester | 50.0 and le 75.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...