异麦芽糖 ≥97.0% (by GC)
供应商: TCI
Synonyms:
6-O-alpha-D-Glucopyranosyl-D-glucose, (2R,3S,4R,5R)-2,3,4,5-Tetrahydroxy-6-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, alpha-1,6-Glucobiose
I0231-1G
I0231-100MG
TCIAI0231-1GEA
0
CNY
TCIAI0231-1G
TCIAI0231-100MG
异麦芽糖 ≥97.0% (by GC)
异麦芽糖
分子式:
C₁₂H₂₂O₁₁ |
MDL编号:
MFCD00065373 CAS号: 499-40-1 EINECS: 207-879-1 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % (total of isomer) |
Specific rotation [a]20/D | +108 to +118 deg (C=0.7, H2O) |
Solubility in Water | almost transparency |
Water | max. 5.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...