Isostearic acid (=2,2,4,8,10,10-Hexamethylundecane-5-carboxylic acid) ≥90.0% (通过GC)

供应商: TCI
Warning

I0184-25G I0184-500G
TCIAI0184-25GEA 0 CNY
TCIAI0184-25G TCIAI0184-500G
Isostearic acid (=2,2,4,8,10,10-Hexamethylundecane-5-carboxylic acid) ≥90.0% (通过GC)
Isostearic acid (=2,2,4,8,10,10-Hexamethylundecane-5-carboxylic acid)
分子式: C₁₈H₃₆O₂
闪点: 165°C
MDL编号: MFCD00135620
CAS号: 54680-48-7

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR