5-Isopropyl-m-xylene ≥85.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
3,5-Dimethylcumene, 3,5-Dimethylisopropylbenzene
I0156-25ML
I0156-500ML
TCIAI0156-25MLEA
0
CNY
TCIAI0156-25ML
TCIAI0156-500ML
5-Isopropyl-m-xylene ≥85.0% (by GC)
5-Isopropyl-m-xylene
分子式:
C₁₁H₁₆ 沸点: 195°C |
MDL编号:
MFCD00026405 CAS号: 4706-90-5 UN号: 3295 ADR: 3,III |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 85.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...