Isonicotinoyl chloride hydrochloride ≥95.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: Pyridine-4-carbonyl Chloride Hydrochloride, 4-Pyridinoyl chloride hydrochloride

I0144-25G I0144-250G
TCIAI0144-25GEA 0 CNY
TCIAI0144-25G TCIAI0144-250G
Isonicotinoyl chloride hydrochloride ≥95.0% (by titrimetric analysis)
Isonicotinoyl chloride hydrochloride
分子式: C₆H₄ClNO
分子量: 178.01 g/mol
MDL编号: MFCD00012830
CAS号: 39178-35-3
EINECS: 254-331-2
UN号: 3261
ADR: 8 II

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 95.0 %
Melting point 159.0 to 162.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR