Isobutylamine hydrochloride ≥99.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
2-Methylpropan-1-amine hydrochloride, 1-Amino-2-methylpropane Hydrochloride
I0096-25G
I0096-500G
TCIAI0096-25GEA
0
CNY
TCIAI0096-25G
TCIAI0096-500G
Isobutylamine hydrochloride ≥99.0% (by titrimetric analysis)
Isobutylamine hydrochloride
分子式:
C₄H₁₁N 分子量: 109.60 g/mol |
MDL编号:
MFCD00060199 CAS号: 5041-09-8 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 99.0 % |
Purity (Argentmetric Titration) | min. 99.0 % |
Melting point | 176.0 to 180.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...