Hydroxychloroquine sulphate ≥98.0% (by HPLC, total nitrogen)

供应商: TCI
Warning

Synonyms: 2-((4-((7-Chloroquinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol sulphate, N4-(7-Chloro-4-quinolyl)-N1-ethyl-N1-(2-hydroxyethyl)-1,4-pentanediamine sulphate, 7-Chloro-4-[4-[N-ethyl-N-(2-hydroxyethyl)amino]-1-methylbutylamino]quinoline Sulphate

H1306-25G H1306-5G
TCIAH1306-25GEA 0 CNY
TCIAH1306-25G TCIAH1306-5G
Hydroxychloroquine sulphate ≥98.0% (by HPLC, total nitrogen)
Hydroxychloroquine sulphate
分子式: C₁₈H₂₆ClN₃O
分子量: 433.95 g/mol
MDL编号: MFCD00078203
CAS号: 747-36-4
EINECS: 212-019-3
Merck Index: 14,04818

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR