N-羟乙基丙烯酰胺 ≥98.0% (by GC, titration analysis) stabilised
供应商: TCI
|
Warning
|
Synonyms:
N-(2-羟乙基)丙烯酰胺, HEAA
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
H1262-25G
H1262-500G
TCIAH1262-25GEA
0
CNY
TCIAH1262-25G
TCIAH1262-500G
N-羟乙基丙烯酰胺 ≥98.0% (by GC, titration analysis) stabilised
N-羟乙基丙烯酰胺
分子式:
C₅H₉NO₂ 分子量: 115.13 g/mol 沸点: 130°C 存储条件: Refrigerator |
MDL编号:
MFCD11110645 CAS号: 7646-67-5 |
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Bromination Method) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...