4-(Hydroxymethyl)cyclohexanol (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC)

供应商: TCI

Synonyms: 4-(Hydroxymethyl)cyclohexanol

H1257-25G H1257-5G
TCIAH1257-25GEA 0 CNY
TCIAH1257-25G TCIAH1257-5G
4-(Hydroxymethyl)cyclohexanol (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC)
4-(Hydroxymethyl)cyclohexanol (cis- and trans- mixture)
分子式: C₇H₁₄O₂
分子量: 130.19 g/mol
沸点: 122°C
MDL编号: MFCD11036248
CAS号: 33893-85-5

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 % (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR