1-[2-(2-羟基乙氧基)乙基]哌嗪 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 2-[2-(1-哌嗪基)乙氧基]乙醇

H1192-25G H1192-5G
TCIAH1192-25GEA 0 CNY
TCIAH1192-25G TCIAH1192-5G
1-[2-(2-羟基乙氧基)乙基]哌嗪 ≥98.0% (by GC, titration analysis)
1-[2-(2-羟基乙氧基)乙基]哌嗪
分子式: C₈H₁₈N₂O₂
沸点: 172°C
MDL编号: MFCD00074931
CAS号: 13349-82-1

现在订购

产品技术规格

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR