1,2-二甲基-3-羟基-4-吡啶酮 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 3-Hydroxy-1,2-dimethylpyridin-4(1H)-one, Deferiprone, 1,2-Dimethyl-3-hydroxy-4-pyridone

H0944-25G H0944-5G
TCIAH0944-25GEA 0 CNY
TCIAH0944-25G TCIAH0944-5G
1,2-二甲基-3-羟基-4-吡啶酮 ≥98.0% (by titrimetric analysis)
1,2-二甲基-3-羟基-4-吡啶酮
分子式: C₇H₉NO₂
分子量: 139.15 g/mol
溶点: 260°C
MDL编号: MFCD00134497
CAS号: 30652-11-0
Merck Index: 14,02859

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Acetic acid almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR