3alpha-Hydroxy-7-oxo-5beta-cholanic acid 97,0 Titrimetric

供应商: TCI

Synonyms: 3-Alpha-Hydroxy-7-oxo-5beta-cholanic acid, 3alpha-Hydroxy-7-keto-5beta-cholanic acid

H0869-25G H0869-5G
TCIAH0869-25GEA 0 CNY
TCIAH0869-25G TCIAH0869-5G
3alpha-Hydroxy-7-oxo-5beta-cholanic acid 97,0 Titrimetric
3alpha-Hydroxy-7-oxo-5beta-cholanic acid
分子式: C₂₄H₃₈O₄
分子量: 390.56 g/mol
MDL编号: MFCD00271393
CAS号: 4651-67-6
EINECS: 225-083-2

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 200.0 to 206.0 deg-C
Specific rotation [a]20/D -27.0 to -31.0 deg (C=2, Ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR