(1S,2S,5S)-(-)-2-羟基-3-蒎烷酮 ≥98.0% (通过GC)
供应商: TCI
H0863-1G
H0863-5G
TCIAH0863-1GEA
0
CNY
TCIAH0863-1G
TCIAH0863-5G
(1S,2S,5S)-(-)-2-羟基-3-蒎烷酮 ≥98.0% (通过GC)
(1S,2S,5S)-(-)-2-羟基-3-蒎烷酮
分子式:
C₁₀H₁₆O₂ |
MDL编号:
MFCD00040556 CAS号: 1845-25-6 |
产品技术规格
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | -32.0 to -38.0 deg (C=0.5, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...