(1R,2R,5R)-(+)-2-羟基-3-蒎烷酮 ≥98.0% (通过GC)

供应商: TCI

H0862-1G H0862-5G
TCIAH0862-1GEA 0 CNY
TCIAH0862-1G TCIAH0862-5G
(1R,2R,5R)-(+)-2-羟基-3-蒎烷酮 ≥98.0% (通过GC)
(1R,2R,5R)-(+)-2-羟基-3-蒎烷酮
分子式: C₁₀H₁₆O₂
MDL编号: MFCD00191522
CAS号: 24047-72-1

现在订购

产品技术规格

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +32.0 to +37.0 deg (C=0.5, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR