Methyl-5-(hydroxymethyl)-1H-imidazole-4-carboxylate ≥98.0% (通过滴定分析)
供应商: TCI
H0840-1G
H0840-5G
TCIAH0840-1GEA
0
CNY
TCIAH0840-1G
TCIAH0840-5G
Methyl-5-(hydroxymethyl)-1H-imidazole-4-carboxylate ≥98.0% (通过滴定分析)
Methyl-5-(hydroxymethyl)-1H-imidazole-4-carboxylate
分子式:
C₆H₈N₂O₃ |
MDL编号:
MFCD00191520 CAS号: 82032-43-7 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...