Methyl-9-hydroxynonanoate ≥95.0% (通过GC)
供应商: TCI
H0835-1G
H0835-5G
TCIAH0835-1GEA
0
CNY
TCIAH0835-1G
TCIAH0835-5G
Methyl-9-hydroxynonanoate ≥95.0% (通过GC)
Methyl-9-hydroxynonanoate
分子式:
C₁₀H₂₀O₃ 沸点: 272°C |
MDL编号:
MFCD00191518 CAS号: 34957-73-8 |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...