4-Methoxyphenyl-4-hydroxybenzoate ≥99.0% (通过GC)
供应商: TCI
|
Warning
|
H0825-25G
H0825-5G
TCIAH0825-25GEA
0
CNY
TCIAH0825-25G
TCIAH0825-5G
4-Methoxyphenyl-4-hydroxybenzoate ≥99.0% (通过GC)
4-Methoxyphenyl-4-hydroxybenzoate
分子式:
C₁₄H₁₂O₄ |
MDL编号:
MFCD00191515 CAS号: 50687-62-2 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 190.0 to 195.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...