Hexaethylene glycol monomethyl ether ≥97.0% (by GC)
供应商: TCI
Synonyms:
mPEG6-Alcohol, 2,5,8,11,14,17-Hexaoxanonadecan-19-ol
H0808-1G
H0808-25G
H0808-5G
TCIAH0808-1GEA
0
CNY
TCIAH0808-1G
TCIAH0808-25G
TCIAH0808-5G
Hexaethylene glycol monomethyl ether ≥97.0% (by GC)
Hexaethylene glycol monomethyl ether
分子式:
C₁₃H₂₈O₇ 分子量: 296.36 g/mol 沸点: 167°C |
MDL编号:
MFCD00045996 CAS号: 23601-40-3 EINECS: 245-775-8 |
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...