Dimethyl(2-hydroxyethyl)phosphonate ≥92.0% (by GC)

供应商: TCI
Warning

Synonyms: (2-Hydroxyethyl)phosphonic Acid Dimethyl Ester

H0779-5G
TCIAH0779-5GEA 0 CNY
TCIAH0779-5G
Dimethyl(2-hydroxyethyl)phosphonate ≥92.0% (by GC)
Dimethyl(2-hydroxyethyl)phosphonate
分子式: C₄H₁₁O₄P
分子量: 154.10 g/mol
沸点: 97°C
闪点: 192°C
MDL编号: MFCD00043376
CAS号: 54731-72-5
EINECS: 259-310-1

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 92.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR