2-(2H-苯并三唑-2-基)-4,6-二叔戊基苯酚 ≥98.0% (by GC)

供应商: TCI
Warning

H0718-25G H0718-500G
TCIAH0718-25GEA 0 CNY
TCIAH0718-25G TCIAH0718-500G
2-(2H-苯并三唑-2-基)-4,6-二叔戊基苯酚 ≥98.0% (by GC)
2-(2H-苯并三唑-2-基)-4,6-二叔戊基苯酚
分子式: C₂₂H₂₉N₃O
MDL编号: MFCD00142737
CAS号: 25973-55-1
EINECS: 247-384-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR