3-Hydroxy-7-methoxy-2-naphthoic acid ≥97.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 3-Hydroxy-7-methoxy-2-naphthalenecarboxylic Acid

H0707-25G H0707-5G
TCIAH0707-25GEA 0 CNY
TCIAH0707-25G TCIAH0707-5G
3-Hydroxy-7-methoxy-2-naphthoic acid ≥97.0% (by HPLC, titration analysis)
3-Hydroxy-7-methoxy-2-naphthoic acid
分子式: C₁₂H₁₀O₄
分子量: 218.21 g/mol
MDL编号: MFCD00059082
CAS号: 143355-56-0

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR