Ethyl heptadecanoate ≥97.0% (by GC)

供应商: TCI

Synonyms: Ethyl Margarate, Heptadecanoic Acid Ethyl Ester, Margaric Acid Ethyl Ester

H0526-25G H0526-5G
TCIAH0526-25GEA 0 CNY
TCIAH0526-25G TCIAH0526-5G
Ethyl heptadecanoate ≥97.0% (by GC)
Ethyl heptadecanoate
分子式: C₁₉H₃₈O₂
分子量: 298.51 g/mol
沸点: 200°C
MDL编号: MFCD00026658
CAS号: 14010-23-2
EINECS: 237-822-6

现在订购

产品技术规格

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR