2-Ethylhexyl-4-hydroxybenzoate ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 4-Hydroxybenzoic Acid Octyl Ester, 4-Hydroxybenzoic Acid 2-Ethylhexyl Ester, Octyl 4-Hydroxybenzoate, 2-Ethylhexylparaben, Octylparaben, 2-Ethylhexyl 4-hydroxybenzoate

H0506-25G H0506-500G
TCIAH0506-25GEA 0 CNY
TCIAH0506-25G TCIAH0506-500G
2-Ethylhexyl-4-hydroxybenzoate ≥98.0% (by HPLC, titration analysis)
2-Ethylhexyl-4-hydroxybenzoate
分子式: C₁₅H₂₂O₃
分子量: 250.34 g/mol
沸点: 270°C
MDL编号: MFCD00016472
CAS号: 5153-25-3
EINECS: 225-925-9

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR