Ethylene glycol monohexyl ether ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
Hexyl Cellosolve, 2-(Hexyloxy)ethanol
H0343-25ML
H0343-500ML
TCIAH0343-25MLEA
0
CNY
TCIAH0343-25ML
TCIAH0343-500ML
Ethylene glycol monohexyl ether ≥98.0% (by GC)
Ethylene glycol monohexyl ether
分子式:
C₈H₁₈O₂ 分子量: 146.23 g/mol 沸点: 205°C 溶点: –45°C 闪点: 102°C |
MDL编号:
MFCD00045997 CAS号: 112-25-4 EINECS: 203-951-1 UN号: 2922 ADR: 8 III |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...