3-Hydroxymyristic acid ≥98.0% (通过GC,滴定分析)

供应商: TCI

H0274-1G H0274-5G
TCIAH0274-1GEA 0 CNY
TCIAH0274-1G TCIAH0274-5G
3-Hydroxymyristic acid ≥98.0% (通过GC,滴定分析)
3-Hydroxymyristic acid
分子式: C₁₄H₂₈O₃
MDL编号: MFCD00059633
CAS号: 1961-72-4

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 78.0 to 81.0 deg-C
Solubility in EtOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR