4-Hydroxy-4'-dimethylaminoazobenzene ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 4-(4-Dimethylaminophenylazo)phenol

H0237-1G H0237-25G
TCIAH0237-1GEA 0 CNY
TCIAH0237-1G TCIAH0237-25G
4-Hydroxy-4'-dimethylaminoazobenzene ≥98.0% (by HPLC, titration analysis)
4-Hydroxy-4'-dimethylaminoazobenzene
分子式: C₁₄H₁₅N₃O
MDL编号: MFCD00059624
CAS号: 2496-15-3

现在订购

产品技术规格

Appearance Orange to Brown to Dark red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR