Histamine diphosphate ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 2-(4-Imidazolyl)ethylamine diphosphate salt, 2-(1H-Imidazol-4-yl)ethan-1-amine bis(phosphate), Histamine phosphate

H0147-1G H0147-5G
TCIAH0147-1GEA 0 CNY
TCIAH0147-1G TCIAH0147-5G
Histamine diphosphate ≥98.0% (by titrimetric analysis)
Histamine diphosphate
分子式: C₅H₉N₃·2H₃PO₄
分子量: 307.14 g/mol
溶点: 133°C
存储条件: Refrigerator
MDL编号: MFCD00064316
CAS号: 51-74-1
EINECS: 200-118-4
Merck Index: 14,04719
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6aa7fbbc7a22086e2f54d78eeb039b4446c7baa1&vid=0f48db86851ce26fcc708c74b45d8bcca193bd55&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water max. 7.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR