Hexamethonium chloride dihydrate ≥99.0% (by titrimetric analysis)

供应商: TCI

Synonyms: N,N,N,N',N',N'-Hexamethyl-1,6-hexanediammonium Dichloride, Hexane-1,6-bis(trimethylammonium Chloride), Hexamethylenebis(trimethylammonium Chloride)

H0090-25G H0090-500G
TCIAH0090-25GEA 0 CNY
TCIAH0090-25G TCIAH0090-500G
Hexamethonium chloride dihydrate ≥99.0% (by titrimetric analysis)
Hexamethonium chloride dihydrate
分子式: C₁₂H₃₀Cl₂N₂·2H₂O
分子量: 273.29 g/mol (anhy)
溶点: 292°C
MDL编号: MFCD00031563
CAS号: 60-25-3
EINECS: 200-465-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=977733ffe6247e4c415d4275a73947017c4d15df&vid=bbd8ed869937090adb197c9b3b3c68d0720241e3&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White powder to crystal
Purity (Argentmetric Titration) min. 99.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water 11.0 to 12.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR