甘油缩甲醛 ≥98.0% (by GC)

供应商: TCI

Synonyms: Glycerol Formal (mixture of 1,3-Dioxan-5-ol and 4-Hydroxymethyldioxolane)

G0244-25ML G0244-500ML
TCIAG0244-25MLEA 0 CNY
TCIAG0244-25ML TCIAG0244-500ML
甘油缩甲醛 ≥98.0% (by GC)
甘油缩甲醛
分子式: C₄H₈O₃
闪点: 93°C
MDL编号: MFCD00003218
CAS号: 99569-11-6
Merck Index: 14,04485

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR