Fmoc-His(Trt)-OH ≥98.0% (by HPLC)

供应商: TCI

Synonyms: Nα-Fmoc-N(im)-三苯甲基-L-组氨酸

F0653-25G F0653-5G
TCIAF0653-25GEA 0 CNY
TCIAF0653-25G TCIAF0653-5G
Fmoc-His(Trt)-OH ≥98.0% (by HPLC)
Fmoc-His(Trt)-OH
分子式: C₄₀H₃₃N₃O₄
分子量: 619.72 g/mol
MDL编号: MFCD00043332
CAS号: 109425-51-6
EINECS: 439-640-4

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 94.0 %
Specific rotation [a]20/D +92.0 to +100.0 deg (C=5, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR