Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride ≥98.0% (by HPLC)

供应商: TCI

Synonyms: Fmoc-Lys-OH.HCl, Nɑ-Fmoc-L-lysine Hydrochloride, Fmoc-Lys-OH·HCl, Nα-Fmoc-L-lysine hydrochloride

F0586-1G F0586-5G
TCIAF0586-1GEA 0 CNY
TCIAF0586-1G TCIAF0586-5G
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride ≥98.0% (by HPLC)
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride
分子式: C₂₁H₂₄N₂O₄
分子量: 404.89 g/mol
MDL编号: MFCD00190889
CAS号: 139262-23-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 94.0 %
Specific rotation [a]20/D -9.0 to -13.0 deg (C=1, DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR