Famotidine ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: 3-(2-Guanidinothiazol-4-ylmethylthio)-N1-sulphamoylpropionamide, 3-(((2-Guanidinothiazol-4-yl)methyl)thio)-N-sulphamoylpropanimidamide

F0530-25G F0530-5G
TCIAF0530-25GEA 0 CNY
TCIAF0530-25G TCIAF0530-5G
Famotidine ≥98.0% (by HPLC, titration analysis)
Famotidine
分子式: C₈H₁₅N₇O₂S₃
分子量: 337.44 g/mol
MDL编号: MFCD00079297
CAS号: 76824-35-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in N,N-DMF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR