Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine-tert-butyl ester ≥98.0% (by HPLC, total nitrogen)

供应商: TCI

Synonyms: Nɑ-Fmoc-L-threonine tert-Butyl Ester, Fmoc-Thr-OtBu, (2S,3R)-tert-Butyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoate, Nα-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine tert-Butyl Ester

F0517-1G F0517-5G
TCIAF0517-1GEA 0 CNY
TCIAF0517-1G TCIAF0517-5G
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine-tert-butyl ester ≥98.0% (by HPLC, total nitrogen)
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine-tert-butyl ester
分子式: C₂₃H₂₇NO₅
分子量: 397.47 g/mol
存储条件: Refrigerator
MDL编号: MFCD08276378
CAS号: 120791-76-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 98.0 to 102.0 deg-C
Specific rotation [a]20/D -4.0 to -11.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR