Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine-tert-butyl ester ≥98.0% (by HPLC, total nitrogen)
供应商: TCI
Synonyms:
Fmoc-Ser-OtBu, (S)-tert-Butyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxypropanoate, Nɑ-Fmoc-L-serine tert-Butyl Ester
F0516-1G
F0516-5G
TCIAF0516-1GEA
0
CNY
TCIAF0516-1G
TCIAF0516-5G
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine-tert-butyl ester ≥98.0% (by HPLC, total nitrogen)
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine-tert-butyl ester
分子式:
C₂₂H₂₅NO₅ 分子量: 383.44 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD01861331 CAS号: 110797-35-8 |
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 123.0 to 127.0 deg-C |
Specific rotation [a]20/D | +6.0 to +9.0 deg (C=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...