4-氟苯甲酰乙酸甲酯 ≥98.0% (by GC)

供应商: TCI

Synonyms: 3-(4-氟苯基)-3-氧丙酸甲酯

F0472-1G F0472-5G
TCIAF0472-1GEA 0 CNY
TCIAF0472-1G TCIAF0472-5G
4-氟苯甲酰乙酸甲酯 ≥98.0% (by GC)
4-氟苯甲酰乙酸甲酯
分子式: C₁₀H₉FO₃
分子量: 196.18 g/mol
沸点: 91°C
MDL编号: MFCD00000355
CAS号: 63131-29-3
EINECS: 263-889-6

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR