N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: N-Fmoc-L-aspartic Acid, Fmoc-Asp-OH

F0452-1G F0452-25G F0452-5G
TCIAF0452-1GEA 0 CNY
TCIAF0452-1G TCIAF0452-25G TCIAF0452-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid ≥98.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid
分子式: C₁₉H₁₇NO₆
分子量: 355.35 g/mol
MDL编号: MFCD00237654
CAS号: 119062-05-4

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -27.0 to -32.0deg (C=1, DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR