Fmoc-Phe(4-NO2)-OH ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: Fmoc-4-硝基-L-苯丙氨酸

F0443-1G F0443-5G
TCIAF0443-1GEA 0 CNY
TCIAF0443-1G TCIAF0443-5G
Fmoc-Phe(4-NO2)-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-Phe(4-NO2)-OH
分子式: C₂₄H₂₀N₂O₆
分子量: 432.43 g/mol
MDL编号: MFCD00057810
CAS号: 95753-55-2

现在订购

产品技术规格

Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -37.0 to -43.0 deg (C=1,DMF)
Solubility in N,N-DMF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR