Ethyl-2-formylpropionate ≥97.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 2-Formylpropionic Acid Ethyl Ester

F0411-1G F0411-5G
TCIAF0411-1GEA 0 CNY
TCIAF0411-1G TCIAF0411-5G
Ethyl-2-formylpropionate ≥97.0% (by titrimetric analysis)
Ethyl-2-formylpropionate
分子式: C₆H₁₀O₃
存储条件: Freezer
MDL编号: MFCD00044012
CAS号: 27772-62-9
UN号: 1993
ADR: 3,III

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR