Fluoromethyl phenyl sulphone ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Fluoromethylsulphonylbenzene, ((Fluoromethyl)sulphonyl)benzene
F0341-1G
F0341-5G
TCIAF0341-1GEA
0
CNY
TCIAF0341-1G
TCIAF0341-5G
Fluoromethyl phenyl sulphone ≥98.0% (by GC)
Fluoromethyl phenyl sulphone
分子式:
C₇H₇FO₂S |
MDL编号:
MFCD00191650 CAS号: 20808-12-2 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 51.0 to 55.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...