5-Formyl-2-thiophenecarboxylic acid ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
5-Formylthenoic Acid, 5-Formylthiophene-2-carboxylic acid
F0336-1G
F0336-5G
TCIAF0336-1GEA
0
CNY
TCIAF0336-1G
TCIAF0336-5G
5-Formyl-2-thiophenecarboxylic acid ≥98.0% (by GC, titration analysis)
5-Formyl-2-thiophenecarboxylic acid
分子式:
C₆H₄O₃S 分子量: 156.16 g/mol |
MDL编号:
MFCD00191437 CAS号: 4565-31-5 |
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...