4-Fluoro-ɑ-methylstyrene ≥98.0% (by GC) stabilised
供应商: TCI
|
Warning
|
Synonyms:
1-Fluoro-4-(1-methylethenyl)benzene, 2-(4-Fluorophenyl)-1-propene
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC)
F0250-25ML
F0250-5ML
TCIAF0250-25MLEA
0
CNY
TCIAF0250-25ML
TCIAF0250-5ML
4-Fluoro-ɑ-methylstyrene ≥98.0% (by GC) stabilised
4-Fluoro-alpha-methylstyrene
分子式:
C₉H₉F 分子量: 136.17 g/mol 沸点: 183°C 闪点: 56°C 存储条件: Refrigerator |
MDL编号:
MFCD00042297 CAS号: 350-40-3 EINECS: 206-501-2 UN号: 1993 ADR: 3 III |
产品技术规格
Appearance | Colorless to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...