3-(2-Fluorophenoxy)benzyl bromide
供应商: TCI
|
Warning
|
Synonyms:
3'-Bromomethyl-2-fluorodiphenyl Ether
F0236-1G
F0236-100MG
TCIAF0236-1GEA
0
CNY
TCIAF0236-1G
TCIAF0236-100MG
3-(2-Fluorophenoxy)benzyl bromide
3-(2-Fluorophenoxy)benzyl bromide
分子式:
C₁₃H₁₀BrFO 分子量: 281.12 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD00059914 CAS号: 242812-04-0 |
产品技术规格
Appearance | Colorless to Orange to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...