2,2'-联糠醛 ≥96.0% (by GC)

供应商: TCI

Synonyms: 2,2-Furoin

F0082-25G F0082-5G
TCIAF0082-25GEA 0 CNY
TCIAF0082-25G TCIAF0082-5G
2,2'-联糠醛 ≥96.0% (by GC)
2,2'-联糠醛
分子式: C₁₀H₈O₄
MDL编号: MFCD00003246
CAS号: 552-86-3
EINECS: 209-024-8

现在订购

产品技术规格

Appearance White to Light red to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 137.0 to 141.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR